missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorophenyl)furfural, 95+%, Thermo Scientific™

Catalog Number p-3212177
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
34035-03-5
C11H7ClO2
206.63
MFCD00195947
ROJGJNINTRCMBL-UHFFFAOYSA-N
5-4-chlorophenyl furan-2-carbaldehyde, 5-4-chlorophenyl-2-furaldehyde, 5-4-chlorophenyl furfural, 5-4-chloro-phenyl-furan-2-carbaldehyde, 2-formyl-5-4-chlorophenyl furan, 2-furancarboxaldehyde, 5-4-chlorophenyl, 5-4-chlorophenyl-2-furancarboxaldehyde, 5-4-chlorophenyl furan-2-carboxaldehyde, pubchem4001, cambridge id 7179578
676184
5-(4-chlorophenyl)furan-2-carbaldehyde
ClC1=CC=C(C=C1)C1=CC=C(O1)C=O
This item is not returnable. View return policy
34035-03-5
C11H7ClO2
206.63
MFCD00195947
ROJGJNINTRCMBL-UHFFFAOYSA-N
5-4-chlorophenyl furan-2-carbaldehyde, 5-4-chlorophenyl-2-furaldehyde, 5-4-chlorophenyl furfural, 5-4-chloro-phenyl-furan-2-carbaldehyde, 2-formyl-5-4-chlorophenyl furan, 2-furancarboxaldehyde, 5-4-chlorophenyl, 5-4-chlorophenyl-2-furancarboxaldehyde, 5-4-chlorophenyl furan-2-carboxaldehyde, pubchem4001, cambridge id 7179578
676184
5-(4-chlorophenyl)furan-2-carbaldehyde
ClC1=CC=C(C=C1)C1=CC=C(O1)C=O
Melting Point 128°C to 131°C
CAS Min % 95.0
Color Brown
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Needles
Assay Percent Range 95% min. (GC)
Formula Weight 206.63
Percent Purity 95+%
Chemical Name or Material 5-(4-Chlorophenyl)furfural
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'