missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Chlorophenyl)-2-furoic acid, 95+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 198.0°C to 201.0°C |
Color | Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 95+% |
Packaging | Glass bottle |
Quantity | 1 g |
Synonym | 5-4-chlorophenyl-2-furoic acid, 5-4-chlorophenyl furan-2-carboxylic acid, 2-furancarboxylic acid, 5-4-chlorophenyl, 5-4-chloro-phenyl-furan-2-carboxylic acid, 2-furancarboxylic acid,5-4-chlorophenyl, acid, 62, acmc-1an70, 5-p-chlorophenyl furoic acid, 5-4-chlorophenyl furoic acid, 5-p-chlorophenyl-2furoic acid |
Solubility Information | Solubility in water: soluble. Other solubilities: ethanol,acetone,dmf,dioxane |
IUPAC Name | 5-(4-chlorophenyl)furan-2-carboxylic acid |
Formula Weight | 222.63 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'