missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-bromophenyl)-3-methyl-1,2,4-oxadiazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 5-4-bromophenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,5-4-bromophenyl-3-methyl, 1,2,4-oxadiazole, 5-4-bromophenyl-3-methyl, 3-methyl-5-4-bromophenyl-1,2,4-oxadiazole, 5-4-bromo-phenyl-3-methyl-1,2,4 oxadiazole |
Formula Weight | 239.07 |
Percent Purity | 97% |
Chemical Name or Material | 5-(4-Bromophenyl)-3-methyl-1, 2, 4-oxadiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'