missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Quantity | 250 mg |
Formula Weight | 268.14 |
Percent Purity | 95% |
Chemical Name or Material | 5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'