missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Isocyanatophenyl)-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Catalog Number p-3586793
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
941716-86-5
C11H9N3O
199.213
MFCD09817502
ZFRFBJVFEQIMLM-UHFFFAOYSA-N
5-3-isocyanatophenyl-1-methyl-1h-pyrazole, 5-3-isocyanatophenyl-1-methylpyrazole, 3-1-methylpyrazol-5-yl benzenisocyanate, 3-1-methyl-1h-pyrazol-5-yl phenylisocyanate
24229624
5-(3-isocyanatophenyl)-1-methylpyrazole
CN1C(=CC=N1)C2=CC(=CC=C2)N=C=O
This item is not returnable. View return policy
941716-86-5
C11H9N3O
199.213
MFCD09817502
ZFRFBJVFEQIMLM-UHFFFAOYSA-N
5-3-isocyanatophenyl-1-methyl-1h-pyrazole, 5-3-isocyanatophenyl-1-methylpyrazole, 3-1-methylpyrazol-5-yl benzenisocyanate, 3-1-methyl-1h-pyrazol-5-yl phenylisocyanate
24229624
5-(3-isocyanatophenyl)-1-methylpyrazole
CN1C(=CC=N1)C2=CC(=CC=C2)N=C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 5-(3-Isocyanatophenyl)-1-methyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'