missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Hydroxyphenyl)-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-3075705
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
690631-98-2
C10H8N2O3
204.185
MFCD05664420
KOWRVFWZCLHEIU-UHFFFAOYSA-N
5-3-hydroxyphenyl-1h-pyrazole-3-carboxylic acid, 5-3-hydroxy-phenyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-3-hydroxyphenyl, 3-3-hydroxyphenyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 5-3-hydroxyphenyl, 5-3-hydroxyphenyl-2h-pyrazole-3-carboxylic acid, 3-3-carboxy-1h-pyrazol-5-yl phenol, 3-carboxy-5-3-hydroxyphenyl-1h-pyrazole, 5-3-hydroxyphenyl pyrazole-3-carboxylic acid
2794658
3-(3-hydroxyphenyl)-1H-pyrazole-5-carboxylic acid
C1=CC(=CC(=C1)O)C2=NNC(=C2)C(=O)O
This item is not returnable. View return policy
690631-98-2
C10H8N2O3
204.185
MFCD05664420
KOWRVFWZCLHEIU-UHFFFAOYSA-N
5-3-hydroxyphenyl-1h-pyrazole-3-carboxylic acid, 5-3-hydroxy-phenyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 5-3-hydroxyphenyl, 3-3-hydroxyphenyl-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 5-3-hydroxyphenyl, 5-3-hydroxyphenyl-2h-pyrazole-3-carboxylic acid, 3-3-carboxy-1h-pyrazol-5-yl phenol, 3-carboxy-5-3-hydroxyphenyl-1h-pyrazole, 5-3-hydroxyphenyl pyrazole-3-carboxylic acid
2794658
3-(3-hydroxyphenyl)-1H-pyrazole-5-carboxylic acid
C1=CC(=CC(=C1)O)C2=NNC(=C2)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.18
Percent Purity 97%
Chemical Name or Material 5-(3-Hydroxyphenyl)-1H-pyrazole-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'