missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Thienyl)thieno[2,3-d]pyrimidin-4(3H)-one, 97%, Thermo Scientific™

Catalog Number p-7039343
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
215928-54-4
C10H6N2OS2
234.291
MFCD00126795
LFUBTAOTPHKKOS-UHFFFAOYSA-N
5-thiophen-2-yl thieno 2,3-d pyrimidin-4-ol, 5-thiophen-2-yl-3h-thieno 2,3-d pyrimidin-4-one, 5-thien-2-ylthieno 2,3-d pyrimidin-4 3h-one, 5-2-thienyl thieno 2,3-d pyrimidin-4 3h-one, 5-thiophen-2-yl thieno 2,3-d pyrimidin-4 3h-one, maybridge3_001755, cambridge id 5319817, thieno 2,3-d pyrimidin-4 1h-one, 5-2-thienyl, thieno 2,3-d pyrimidin-4 3h-one, 5-2-thienyl, 5-2-thienyl-3-hydrothiopheno 2,3-d pyrimidin-4-one
665387
5-thiophen-2-yl-3H-thieno[2,3-d]pyrimidin-4-one
C1=CSC(=C1)C2=CSC3=C2C(=O)NC=N3
This item is not returnable. View return policy
215928-54-4
C10H6N2OS2
234.291
MFCD00126795
LFUBTAOTPHKKOS-UHFFFAOYSA-N
5-thiophen-2-yl thieno 2,3-d pyrimidin-4-ol, 5-thiophen-2-yl-3h-thieno 2,3-d pyrimidin-4-one, 5-thien-2-ylthieno 2,3-d pyrimidin-4 3h-one, 5-2-thienyl thieno 2,3-d pyrimidin-4 3h-one, 5-thiophen-2-yl thieno 2,3-d pyrimidin-4 3h-one, maybridge3_001755, cambridge id 5319817, thieno 2,3-d pyrimidin-4 1h-one, 5-2-thienyl, thieno 2,3-d pyrimidin-4 3h-one, 5-2-thienyl, 5-2-thienyl-3-hydrothiopheno 2,3-d pyrimidin-4-one
665387
5-thiophen-2-yl-3H-thieno[2,3-d]pyrimidin-4-one
C1=CSC(=C1)C2=CSC3=C2C(=O)NC=N3

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 234.29
Percent Purity 97%
Chemical Name or Material 5-(2-Thienyl)thieno[2,3-d]pyrimidin-4(3H)-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'