5-(2-thienyl)-1,3-oxazole, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: SEW02560EA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 70380-70-0 Weight: 0.06000kg


Packaging Amber Glass Bottle
Quantity 10g

Product Code. 10526603

Quantity Price
1 £ 187.0 / 10g
Estimated Shipment date
from Supplier 03-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 5-(2-Thienyl)-1, 3-oxazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H5NOS
Formula Weight 151.19

Specification

Formula Weight 151.19
Quantity 10g
Chemical Name or Material 5-(2-Thienyl)-1, 3-oxazole
Assay 95%
CAS 70380-70-0
Molecular Formula C7H5NOS
Assay Percent Range 95%
Packaging Amber Glass Bottle
Documentation

Technical documents