missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-Pyridinylsulfanyl)-2-furaldehyde, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 205.24 |
Percent Purity | 95% |
Chemical Name or Material | 5-(2-Pyridinylsulfanyl)-2-furaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'