missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde, 90%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 90.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 212.27 |
Percent Purity | 90% |
Chemical Name or Material | 5-(2-Phenyleth-1-ynyl)thiophene-2-carbaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'