missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Phenyleth-1-ynyl)-2-furoic acid, Tech., Thermo Scientific™

Catalog Number p-2702221
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
130423-83-5
C13H8O3
212.20
MFCD00541949
HFYACYZCOUYIFR-UHFFFAOYSA-N
5-phenylethynyl furan-2-carboxylic acid, 5-2-phenyleth-1-ynyl-2-furoic acid, 5-2-phenylethynyl furan-2-carboxylic acid, 5-phenylethynyl-2-furoic acid, 2-furancarboxylic acid,5-2-phenylethynyl, maybridge1_008982, acmc-20a65w, 5-phenylethynyl-2-furoic acid, tech, 5-phenylethynyl-furan 2 carboxylic acid
2737138
5-(2-phenylethynyl)furan-2-carboxylic acid
OC(=O)C1=CC=C(O1)C#CC1=CC=CC=C1
This item is not returnable. View return policy
130423-83-5
C13H8O3
212.20
MFCD00541949
HFYACYZCOUYIFR-UHFFFAOYSA-N
5-phenylethynyl furan-2-carboxylic acid, 5-2-phenyleth-1-ynyl-2-furoic acid, 5-2-phenylethynyl furan-2-carboxylic acid, 5-phenylethynyl-2-furoic acid, 2-furancarboxylic acid,5-2-phenylethynyl, maybridge1_008982, acmc-20a65w, 5-phenylethynyl-2-furoic acid, tech, 5-phenylethynyl-furan 2 carboxylic acid
2737138
5-(2-phenylethynyl)furan-2-carboxylic acid
OC(=O)C1=CC=C(O1)C#CC1=CC=CC=C1
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.2
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-(2-Phenyleth-1-ynyl)-2-furoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'