missing translation for 'onlineSavingsMsg'
Learn More

[5-(2-Methyl-1,3-thiazol-4-yl)-3-isoxazolyl]methanol, 97%, Thermo Scientific™

Catalog Number 10664745
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 388088-79-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H8N2O2S
Linear Formula Yes
MDL Number MFCD03086111
Quantity 1 g
Synonym 5-2-methyl-1,3-thiazol-4-yl-3-isoxazolyl methanol, 5-2-methyl-1,3-thiazol-4-yl-1,2-oxazol-3-yl methanol, 5-2-methylthiazol-4-yl isoxazol-3-yl methanol, 5-2-methyl-1,3-thiazol-4-yl isoxazol-3-yl methanol, 5-2-methyl-1,3-thiazol-4-yl isoxazol-3-yl methan-1-ol
InChI Key ANQHCDZMYKOBKX-UHFFFAOYSA-N
SMILES CC1=NC(=CS1)C2=CC(=NO2)CO
IUPAC Name [5-(2-methyl-1,3-thiazol-4-yl)-1,2-oxazol-3-yl]methanol
Molecular Weight (g/mol) 196.224
PubChem CID 2776410
Formula Weight 196.23
Percent Purity 97%
Chemical Name or Material [5-(2-Methyl-1, 3-thiazol-4-yl)-3-isoxazolyl]methanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'