missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[5-(2-Methyl-1,3-thiazol-4-yl)-2-thienyl]methanol 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Assay Percent Range | 95% |
Quantity | 1 g |
Formula Weight | 211.31 |
Percent Purity | 95% |
Chemical Name or Material | [5-(2-Methyl-1, 3-thiazol-4-yl)-2-thienyl]methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'