missing translation for 'onlineSavingsMsg'
Learn More

[5-(2-Furyl)-1H-pyrazol-3-yl]methanol, 97%, Thermo Scientific™

Catalog Number 10585953
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
84978-67-6
C8H8N2O2
164.164
NBADTAIDQSLLHA-UHFFFAOYSA-N
5-2-furyl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-1h-pyrazol-3-yl methanol, 3-furan-2-yl-1h-pyrazol-5-yl methanol, 5-2-furyl pyrazol-3-yl methan-1-ol, 1h-pyrazole-3-methanol,5-2-furanyl, 5-2-furyl-1h-pyrazol-4-yl methanol, 5-fur-2-yl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-2h-pyrazol-3-yl methanol
7060534
[3-(furan-2-yl)-1H-pyrazol-5-yl]methanol
C1=COC(=C1)C2=NNC(=C2)CO
This item is not returnable. View return policy
84978-67-6
C8H8N2O2
164.164
NBADTAIDQSLLHA-UHFFFAOYSA-N
5-2-furyl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-1h-pyrazol-3-yl methanol, 3-furan-2-yl-1h-pyrazol-5-yl methanol, 5-2-furyl pyrazol-3-yl methan-1-ol, 1h-pyrazole-3-methanol,5-2-furanyl, 5-2-furyl-1h-pyrazol-4-yl methanol, 5-fur-2-yl-1h-pyrazol-3-yl methanol, 5-furan-2-yl-2h-pyrazol-3-yl methanol
7060534
[3-(furan-2-yl)-1H-pyrazol-5-yl]methanol
C1=COC(=C1)C2=NNC(=C2)CO
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material [5-(2-Furyl)-1H-pyrazol-3-yl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'