missing translation for 'onlineSavingsMsg'
Learn More

5-[2-(Ethylamino)ethylcarbamoyl]-2-fluorobenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15411266
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2096332-79-3
C11H16BFN2O3
254.07
MFCD20040204
UBDRCBYERBTPRG-UHFFFAOYSA-N
5-2-ethylamino ethylcarbamoyl-2-fluorobenzeneboronic acid
73995842
This item is not returnable. View return policy
2096332-79-3
C11H16BFN2O3
254.07
MFCD20040204
UBDRCBYERBTPRG-UHFFFAOYSA-N
5-2-ethylamino ethylcarbamoyl-2-fluorobenzeneboronic acid
73995842

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 254.07
Percent Purity 97%
Chemical Name or Material 5-[2-(Ethylamino)ethylcarbamoyl]-2-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'