missing translation for 'onlineSavingsMsg'
Learn More

5-(2,5-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%, Thermo Scientific™

Catalog Number 15401216
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
2096335-17-8
C13H9BCl2FNO3
327.93
MFCD20040191
NRCZOQMPVBOVGS-UHFFFAOYSA-N
5-2,5-dichlorophenylcarbamoyl-2-fluorobenzeneboronic acid
73995818
This item is not returnable. View return policy
2096335-17-8
C13H9BCl2FNO3
327.93
MFCD20040191
NRCZOQMPVBOVGS-UHFFFAOYSA-N
5-2,5-dichlorophenylcarbamoyl-2-fluorobenzeneboronic acid
73995818

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 327.93
Percent Purity 97%
Chemical Name or Material 5-(2,5-Dichlorophenylcarbamoyl)-2-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'