missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes

Measure, mix and pour using 5.1 Borosilicate Glass Disposable Test Tubes. Tubes are rimmed and are available in various lengths.

Brand:  DWK Life Sciences A510010000521

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11872363

  • £20.95 / Pack of 100
Currently Unavailable: We have no information when this product will be available
View alternative products

Description

Description

  • Disposable
  • Clear borosilicate glass
  • Round bottom
  • Rimmed
Specifications

Specifications

Test Tube
Round bottom
1Each
10 mm
100 mm
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers