missing translation for 'onlineSavingsMsg'
Learn More

(4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%, ACROS Organics™

Brand:  Acros Organics 337860010

Code : 29

Additional Details : CAS Number : 82938-50-9 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10787972

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

(4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
82938-50-9
100.0
C10H19NO3Si
MFCD01862126
99%
98.5
99%
229.35

Specifications

(4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
82938-50-9
100.0
135.0° to 137.0°C
1g
C10H19NO3Si
MFCD01862126
99%
98.5
Glass bottle
99%
99%
229.35
Videos
SDS