(4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 337862500

250MG (4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid, 99%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 82938-50-9 Weight: 0.05025kg


Packaging Glass bottle
Quantity 250mg

Product Code. 10516071

Quantity Price
1 £ 55.2 / 250mg
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material (4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
Name Note 99%
CAS 82938-50-9
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C10H19NO3Si
Formula Weight 229.35
MDL Number MFCD01862126

Specification

Formula Weight 229.35
Quantity 250mg
CAS Min % 98.5
CAS 82938-50-9
Assay Percent Range 99%
MDL Number MFCD01862126
Packaging Glass bottle
Name Note 99%
Molecular Formula C10H19NO3Si
CAS Max % 100.0
Chemical Name or Material (4S)-N-(tert-Butyldimethylsilyl)azetidin-2-one-4-carboxylic acid
TSCA TSCA
Documentation

Technical documents