missing translation for 'onlineSavingsMsg'
Learn More

(4S)-(-)-4,5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole, 98%, ACROS Organics™

Brand:  Acros Organics 353735000

Code : 29

Additional Details : CAS Number : 148461-14-7 Weight : 0.05050kg


Quantity 500mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10733085

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

(4S)-(-)-4, 5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole
97.5
≥97.5%
C24H24NOP
MFCD03427599
148461-14-7
100.0
98%
373.43

Specifications

(4S)-(-)-4, 5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole
148461-14-7
100.0
Glass bottle
Crystalline Powder
98%
White
MFCD03427599
500mg
97.5
Authentic
-43° to -46° (20°C, 589nm) (C=1, CHCl3)
≥97.5%
C24H24NOP
373.43
Videos
SDS