missing translation for 'onlineSavingsMsg'
Learn More

(4R)-(+)-4-Isopropyl-2-oxazolidinone, 98%, ACROS Organics™

Brand:  Acros Organics 297850250

Code : 29

Additional Details : CAS Number : 95530-58-8 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10581734

  • £1166.50 / 25g


Estimated Shipment
20-03-2018
Add to basket

Specifications

Chemical Identifiers

(4R)-(+)-4-Isopropyl-2-oxazolidinone
97.5
≥97.5%
C6H11NO2
MFCD00075172
95530-58-8
100.0
98%
129.16

Specifications

(4R)-(+)-4-Isopropyl-2-oxazolidinone
White
25g
97.5
Crystalline Powder
≥97.5%
C6H11NO2
MFCD00075172
+16° to +20° (20°C, 589nm) (c=1, ethanol)
−18° (20°C c=1,ethanol)
70° to 74°C
95530-58-8
100.0
Authentic
98%
129.16
Glass bottle
Videos
SDS