missing translation for 'onlineSavingsMsg'
Learn More

(4R)-(+)-4-Isopropyl-2-oxazolidinone, 98%, Thermo Scientific Chemicals

Catalog Number 10653742
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
95530-58-8
C6H11NO2
129.16
MFCD00075172
YBUPWRYTXGAWJX-YFKPBYRVSA-N
r-4-isopropyl-2-oxazolidinone, r-4-isopropyloxazolidin-2-one, r-+-4-isopropyl-2-oxazolidinone, 4r-+-4-isopropyl-2-oxazolidinone, 4r-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4r, 4r-4-propan-2-yl-1,3-oxazolidin-2-one, pubchem8176, r-isopropyloxazolidinone
641505
(4R)-4-propan-2-yl-1,3-oxazolidin-2-one
CC(C)[C@@H]1COC(=O)N1
This item is not returnable. View return policy
95530-58-8
C6H11NO2
129.16
MFCD00075172
YBUPWRYTXGAWJX-YFKPBYRVSA-N
r-4-isopropyl-2-oxazolidinone, r-4-isopropyloxazolidin-2-one, r-+-4-isopropyl-2-oxazolidinone, 4r-+-4-isopropyl-2-oxazolidinone, 4r-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4r, 4r-4-propan-2-yl-1,3-oxazolidin-2-one, pubchem8176, r-isopropyloxazolidinone
641505
(4R)-4-propan-2-yl-1,3-oxazolidin-2-one
CC(C)[C@@H]1COC(=O)N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70°C to 73°C
CAS Min % 97.5
CAS Max % 100.0
Color White
Assay Percent Range 97.5% min. (GC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water
Specific Rotation -18
Formula Weight 129.16
Percent Purity 98%
Specific Rotation Condition −18° (20°C c=1,ethanol)
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material (4R)-(+)-4-Isopropyl-2-oxazolidinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'