missing translation for 'onlineSavingsMsg'
Learn More

4F 4PP oxalate, Tocris Bioscience™

Selective 5-HT2A antagonist

Brand:  Tocris (Bio-Techne) 052350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13467683

  • £401.31 / 50mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A selective 5-HT2A antagonist with almost as high affinity (Ki = 5.3 nM) as ketanserin but with a much lower affinity for 5-HT2C sites (Ki = 620 nM).
Specifications

Chemical Identifiers

144734-36-1
429.488
4f 4pp oxalate, 4-4-fluorobenzoyl-1-4-phenylbutyl piperidine oxalate, 4-fluorophenyl 1-4-phenylbutyl piperidin-4-yl methanone ethanedioate
(4-fluorophenyl)-[1-(4-phenylbutyl)piperidin-4-yl]methanone;oxalic acid
C24H28FNO5
VUJYJCRJPFMHEM-UHFFFAOYSA-N
24745966
C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCCCC3=CC=CC=C3.C(=O)(C(=O)O)O

Specifications

50mg
144734-36-1
4f 4pp oxalate, 4-4-fluorobenzoyl-1-4-phenylbutyl piperidine oxalate, 4-fluorophenyl 1-4-phenylbutyl piperidin-4-yl methanone ethanedioate
C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCCCC3=CC=CC=C3.C(=O)(C(=O)O)O
429.488
429.49
4F 4PP oxalate
C24H28FNO5
VUJYJCRJPFMHEM-UHFFFAOYSA-N
(4-fluorophenyl)-[1-(4-phenylbutyl)piperidin-4-yl]methanone;oxalic acid
24745966
>99%
Videos
Documents