Learn More
Tocris Bioscience™ 4E1RCat
Inhibits eIF4F subunit interaction; inhibits protein translation
Brand:  Tocris Bioscience™ 421550
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13434374
  • £633.57 / 50mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Inhibitor of protein translation; blocks eIF4E:eIF4G and eIF4E:4E-BP1 interaction. Prevents assembly of the eIF4F complex and inhibits cap-dependent translation (IC50 ∽4 μM). Exhibits chemosensitizing properties.
Specifications
Specifications
Protein-synthesizing GTPase Compounds
>98
50mg
4E1RCat
478.45