missing translation for 'onlineSavingsMsg'
Learn More

QIAGEN QIAcard FTA PlantSaver

For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.

Brand:  QIAGEN WB120065

Code : 69

Additional Details : CAS Number : 9004-34-6 Weight : 0.47000kg

Product Code. 11994505

  • £967.00 / Pack of 100
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.

The 4 spot area QIAcard FTA PlantSaver format provides a laminated flap that allows you to vigorously crush plant or insect samples into the FTA matrix without damaging the FTA card.

Features:

  • Simply pressing plant samples onto the FTA matrix or applying homogenate captures and stabilizes the DNA.
  • Store and collect at room and high temperatures for convenient collection in the field.
  • Smaller sample size: Only young leaves are required, reducing growing time and speeding research.
  • Fast purification in 30 minutes: PCR-ready DNA can be purified on FTA in three simple steps, all in a single tube at room temperature.

Applications:

  • Analyze plant DNA by PCR-based assays
  • Detection of Genetically Modified (GM) organisms
  • Marker selected breeding
  • Varietal identification
  • Phylogeny analysis
  • Amplification of low copy number loci
  • Invader™ assays
  • Multiple displacement amplification
  • Transgenic identification

Analysis:

  • Plant tissue is physically crushed on the card and the DNA binds to the matrix.
  • The chemical coating on the FTA card protects the DNA from degradation and allows the cards to be stored at room temperature for extended periods of time.
  • To prepare the sample for PCR, the matrix is washed with two nontoxic aqueous buffers.
  • The DNA remains bound to the matrix throughout purification and a small disk (1.2 or 2 mm in diameter) of the matrix provides enough template for PCR analysis.
Specifications

Specifications

FTA PlantSaver card with 4 sample areas per card
Plants, Biological
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology
FTA PlantSaver Card
Card
Indicating
4
100 Pk.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers