missing translation for 'onlineSavingsMsg'
Learn More

Retsch™ 400 dia. x 65mmH Stainless Steel Test Sieve, ISO Certified

Brand:  Retsch™ 60.166.000150

Code : A0

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 10630462

  • £229.33 / Each


Estimated Shipment
29-06-2018
Add to basket

Specifications

Specifications

For separation, fractioning, particle size determination
Stainless Steel
150μm
ISO 3310/1
Shaker AS 450
Stainless Steel
1Each
400mm
Test Sieve
Stainless Steel
65mm
Videos