missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 400 dia. x 65mmH Stainless Steel Test Sieve, ISO Certified

Brand:  RETSCH 60.166.000160

Code : NEW

Additional Details : Weight : 1.60000kg

 View more versions of this product

Product Code. 10600292

  • £273.00 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Stainless Steel
Stainless Steel
1Each
Test Sieve
Stainless Steel
65mm
For separation, fractioning, particle size determination
400 mm
160μm
ISO 3310/1
Shaker AS 450
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers