missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylphenol, 95%, 10% solution in propylene glycol, ACROS Organics™

Brand:  Acros Organics 434041000

Code : 29

Additional Details : CAS Number : 2628-17-3 Weight : 0.15390kg


Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Product Code. 11914671

  • £571.20 / 100mL


Estimated Shipment
14-02-2018
Add to basket


Specifications

Chemical Identifiers

4-Vinylphenol
9.0
95%
120.16
57-55-6
11.0
C8H8O
4-Ethenylphenol

Specifications

4-Vinylphenol
9.0
Authentic
1.4420 to 1.4520 (20°C, 589nm)
95%
120.16
4-Ethenylphenol
100mL
57-55-6
11.0
Glass bottle
Liquid
C8H8O
1.039g/mL
Clear colorless to orange
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Vinylphenol, 10% solution in propylene glycol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS