missing translation for 'onlineSavingsMsg'
Learn More

4-Trimethylsilyl-3-butyn-1-ol, 98%, Thermo Scientific Chemicals

Catalog Number 10065463
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2117-12-6
C7H14OSi
142.27
MFCD00190217
VSTMVAFCUIGHQI-UHFFFAOYSA-N
4-trimethylsilyl-3-butyn-1-ol, 4-trimethylsilyl but-3-yn-1-ol, 3-butyn-1-ol, 4-trimethylsilyl, 4-trimethylsilyl-3-butyne-1-ol, 1-trimethylsilyl-1-butyn-4-ol, 3-butyn-1-ol,4-trimethylsilyl
2760827
4-trimethylsilylbut-3-yn-1-ol
C[Si](C)(C)C#CCCO
This item is not returnable. View return policy
2117-12-6
C7H14OSi
142.27
MFCD00190217
VSTMVAFCUIGHQI-UHFFFAOYSA-N
4-trimethylsilyl-3-butyn-1-ol, 4-trimethylsilyl but-3-yn-1-ol, 3-butyn-1-ol, 4-trimethylsilyl, 4-trimethylsilyl-3-butyne-1-ol, 1-trimethylsilyl-1-butyn-4-ol, 3-butyn-1-ol,4-trimethylsilyl
2760827
4-trimethylsilylbut-3-yn-1-ol
C[Si](C)(C)C#CCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Black-Brown to Gray
Density 0.8540g/mL
Boiling Point 97.0°C to 98.0°C (24.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3SiC≡C(CH2)2OH
Refractive Index 1.4540 to 1.4560 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.854
Formula Weight 142.27
Percent Purity 98%
Physical Form Solution
Chemical Name or Material 4-Trimethylsilyl-3-butyn-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Trimethylsilyl-3-butyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'