missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)phenylhydrazine, 96%, Thermo Scientific Chemicals

Catalog Number 10516504
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
368-90-1
C7H7F3N2
176.14
MFCD00042508
DBNLGTYGKCMLLR-UHFFFAOYSA-N
4-trifluoromethyl phenylhydrazine, 4-trifluoromethyl phenyl hydrazine, 4-trifluoromethylphenylhydrazine, p-trifluoromethylphenylhydrazine, hydrazine, 4-trifluoromethyl phenyl, p-trifluoromethyl phenylhydrazine, 4-trifluoromethyl-phenylhydrazine, 4-trifluoromethyl-phenyl-hydrazine
136200
[4-(trifluoromethyl)phenyl]hydrazine
NNC1=CC=C(C=C1)C(F)(F)F
This item is not returnable. View return policy
368-90-1
C7H7F3N2
176.14
MFCD00042508
DBNLGTYGKCMLLR-UHFFFAOYSA-N
4-trifluoromethyl phenylhydrazine, 4-trifluoromethyl phenyl hydrazine, 4-trifluoromethylphenylhydrazine, p-trifluoromethylphenylhydrazine, hydrazine, 4-trifluoromethyl phenyl, p-trifluoromethyl phenylhydrazine, 4-trifluoromethyl-phenylhydrazine, 4-trifluoromethyl-phenyl-hydrazine
136200
[4-(trifluoromethyl)phenyl]hydrazine
NNC1=CC=C(C=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 59°C to 64°C
Color Yellow to Brown
Boiling Point 118°C to 122°C (17.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CF3C6H4NHNH2
Quantity 5 g
Formula Weight 176.14
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-(Trifluoromethyl)phenylhydrazine
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethyl)phenylhydrazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'