missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)nicotinonitrile, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Especificaciones
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 172.11 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Trifluoromethyl)nicotinonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'