missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)nicotinonitrile, ≥95%, Maybridge

Brand:  Maybridge RF04123DE

Code : 31

Additional Details : CAS Number : 13600-43-6 Weight : 0.05500kg Transport : UN number : 3276 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 10639505

  • £170.00 / 5g


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Chemical Identifiers

4-(Trifluoromethyl)nicotinonitrile
97.0
97%
172.11
13600-43-6
97%
C7H3F3N2
MFCD00153107

Specifications

4-(Trifluoromethyl)nicotinonitrile
13600-43-6
97%
C7H3F3N2
MFCD00153107
5g
97.0
97%
172.11
Videos
SDS