missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific™

Catalog Number p-2700972
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
158063-66-2
C7H4F3NO2
191.11
MFCD00082626
LMRJHNFECNKDKH-UHFFFAOYSA-N
4-trifluoromethyl nicotinic acid, 4-trifluoromethyl pyridine-3-carboxylic acid, tfna, 3-pyridinecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-nicotinic acid, 4-trifluoromethylnicotinic acid, 3-carboxy-4-trifluoromethyl pyridine, 4-trifluoromethyl-3-pyridinecarboxylic acid, maybridge1_006267
2777549
4-(trifluoromethyl)pyridine-3-carboxylic acid
OC(=O)C1=C(C=CN=C1)C(F)(F)F
This item is not returnable. View return policy
158063-66-2
C7H4F3NO2
191.11
MFCD00082626
LMRJHNFECNKDKH-UHFFFAOYSA-N
4-trifluoromethyl nicotinic acid, 4-trifluoromethyl pyridine-3-carboxylic acid, tfna, 3-pyridinecarboxylic acid, 4-trifluoromethyl, 4-trifluoromethyl-nicotinic acid, 4-trifluoromethylnicotinic acid, 3-carboxy-4-trifluoromethyl pyridine, 4-trifluoromethyl-3-pyridinecarboxylic acid, maybridge1_006267
2777549
4-(trifluoromethyl)pyridine-3-carboxylic acid
OC(=O)C1=C(C=CN=C1)C(F)(F)F
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 191.11
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethyl)nicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'