missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)imidazole, 98%, Thermo Scientific Chemicals

Catalog Number p-7055982
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
33468-69-8
C4H3F3N2
136.077
MFCD08458860
DFLGRTIPTPCKPJ-UHFFFAOYSA-N
4-trifluoromethyl-1h-imidazole, 4-trifluoromethylimidazole, 4-trifluoromethyl imidazole, 5-trifluoromethyl-1h-imidazole, 4 5-trifluoromethyl imidazole, 1h-imidazole, 4-trifluoromethyl, imidazole, 4-trifluoromethyl, pubchem19080, acmc-1ailu
585891
5-(trifluoromethyl)-1H-imidazole
C1=C(NC=N1)C(F)(F)F
This item is not returnable. View return policy
33468-69-8
C4H3F3N2
136.077
MFCD08458860
DFLGRTIPTPCKPJ-UHFFFAOYSA-N
4-trifluoromethyl-1h-imidazole, 4-trifluoromethylimidazole, 4-trifluoromethyl imidazole, 5-trifluoromethyl-1h-imidazole, 4 5-trifluoromethyl imidazole, 1h-imidazole, 4-trifluoromethyl, imidazole, 4-trifluoromethyl, pubchem19080, acmc-1ailu
585891
5-(trifluoromethyl)-1H-imidazole
C1=C(NC=N1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 148°C to 150°C
Quantity 250 mg
Formula Weight 136.08
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(Trifluoromethyl)imidazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'