missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)hydrocinnamic acid, 95%, ACROS Organics™

Brand:  Acros Organics 397380050

Code : 29

Additional Details : CAS Number : 53473-36-2 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10663215

  • £75.80 / 5g


Estimated Shipment
20-06-2018
Add to basket


Specifications

Chemical Identifiers

4-(Trifluoromethyl)hydrocinnamic acid
94.0
95%
C10H9F3O2
218.17
53473-36-2
100.0
95%
COOH(CH2)2C6H4CF3
MFCD00674032

Specifications

4-(Trifluoromethyl)hydrocinnamic acid
53473-36-2
Glass bottle
106.0° to 110.0°C
5g
95%
C10H9F3O2
218.17
Authentic
94.0
100.0
White to light brown
Crystalline Powder or Solid
95%
COOH(CH2)2C6H4CF3
MFCD00674032
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-(Trifluoromethyl)hydrocinnamic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS