missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Trifluoromethyl-D-phenylalanine, 98%

Brand:  Alfa Aesar™ H64132.06

Code : NEW

Additional Details : CAS Number : 114872-99-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15421199

  • £470.64 / 5g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

4-Trifluoromethyl-D-phenylalanine
233.19
(R)-2-Amino-3-[4-(trifluoromethyl)phenyl]propionic acid
114872-99-0
MFCD01860876

Specifications

4-Trifluoromethyl-D-phenylalanine
Powder
233.19
(R)-2-Amino-3-[4-(trifluoromethyl)phenyl]propionic acid
5g
114872-99-0
98%
MFCD01860876
White
Videos
SDS