missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethyl)cyclohexan-1-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CF3C6H9(=O) |
Quantity | 1 g |
Synonym | 4-trifluoromethyl cyclohexanone, 4-trifluoromethyl cyclohexan-1-one, 4-trifluoromethylcyclohexanone, cyclohexanone, 4-trifluoromethyl, 4-trifluoromethyl-1-cyclohexanone, pubchem16025, 4-trifluoromethyl-cyclohexanone, cyclohexanone,4-trifluoromethyl, 1-oxo-4-trifluoromethyl cyclohexane |
Formula Weight | 166.14 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Trifluoromethyl)cyclohexan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'