missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Trifluoromethyl)benzamidoxime, 97%

Brand:  Alfa Aesar™ H58755.09

Code : NEW

Additional Details : CAS Number : 22179-86-8 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 15404978

  • £664.62 / 10g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

4-(Trifluoromethyl)benzamidoxime
22179-86-8
204.15
MFCD00053042

Specifications

4-(Trifluoromethyl)benzamidoxime
125° to 130°C
22179-86-8
97%
MFCD00053042
Pale brown
10g
Solid
204.15
Videos
SDS