Learn More
Alfa Aesar™ 4-(Trifluoromethyl)anisole, 98%
Brand: Alfa Aesar™ H64187.MD
Code : NEW
Additional Details : CAS Number : 402-52-8 Weight : 0.00025kg Transport : UN number : 1993 Chem class : 1 Pack group : III
Quantity | 250mg |
---|
Chemical Identifiers
4-(Trifluoromethyl)anisole | |
176.14 | |
UN1993 |
402-52-8 | |
MFCD03094456 | |
1-Methoxy-4-(trifluoromethyl)benzene |
Specifications
4-(Trifluoromethyl)anisole | |
169°C (754mmHg) | |
−9°C | |
402-52-8 | |
Liquid | |
176.14 | |
UN1993 |
1.245g/mL | |
Colorless to pale yellow | |
250mg | |
1.4450 | |
98% | |
MFCD03094456 | |
1-Methoxy-4-(trifluoromethyl)benzene |