missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Trifluoromethyl)anisole, 98%

Brand:  Alfa Aesar™ H64187.06

Code : NEW

Additional Details : CAS Number : 402-52-8 Weight : 0.00500kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15461279

  • £136.70 / 5g


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Specifications

Chemical Identifiers

4-(Trifluoromethyl)anisole
176.14
UN1993
402-52-8
MFCD03094456
1-Methoxy-4-(trifluoromethyl)benzene

Specifications

4-(Trifluoromethyl)anisole
169°C (754mmHg)
−9°C
402-52-8
Liquid
176.14
UN1993
1.245g/mL
Colorless to pale yellow
5g
1.4450
98%
MFCD03094456
1-Methoxy-4-(trifluoromethyl)benzene
Videos
SDS