missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethyl)acetophenone, 99%, Thermo Scientific Chemicals

Catalog Number 10510984
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
709-63-7
C9H7F3O
188.15
MFCD00000401
HHAISVSEJFEWBZ-UHFFFAOYSA-N
4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl
69731
1-[4-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=CC=C(C=C1)C(F)(F)F
This item is not returnable. View return policy
709-63-7
C9H7F3O
188.15
MFCD00000401
HHAISVSEJFEWBZ-UHFFFAOYSA-N
4'-trifluoromethyl acetophenone, 1-4-trifluoromethyl phenyl ethanone, 1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethylacetophenone, 4-acetylbenzotrifluoride, 4-trifluoromethyl acetophenone, p-trifluoromethylacetophenone, 4'-trifluoromethylacetophenone, ethanone, 1-4-trifluoromethyl phenyl
69731
1-[4-(trifluoromethyl)phenyl]ethanone
CC(=O)C1=CC=C(C=C1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 30°C to 33°C
Color White to Yellow
Density 0.9200g/mL
Boiling Point 79°C to 80°C (8.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3C6H4COCH3
Refractive Index 1.4604
Quantity 25 g
Specific Gravity 0.92
Formula Weight 188.15
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 4'-(Trifluoromethyl)acetophenone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4'-(Trifluoromethyl)acetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'