missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)thiophenol, 95%, Thermo Scientific Chemicals

Catalog Number p-7047313
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
169685-29-4
C7H5F3OS
194.171
MFCD00052312
JHVNSRJPBXPZJU-UHFFFAOYSA-N
4-trifluoromethoxy thiophenol, 4-trifluoromethoxy benzenethiol, 4-trifluoromethoxy benzene-1-thiol, 4-trifluoromethoxythiophenol, benzenethiol, 4-trifluoromethoxy, pubchem15268, acmc-1bxy9, 4-trifluoromethoxy-thiophenol, p-trifluoromethoxybenzenethiol
2777360
4-(trifluoromethoxy)benzenethiol
C1=CC(=CC=C1OC(F)(F)F)S
This item is not returnable. View return policy
169685-29-4
C7H5F3OS
194.171
MFCD00052312
JHVNSRJPBXPZJU-UHFFFAOYSA-N
4-trifluoromethoxy thiophenol, 4-trifluoromethoxy benzenethiol, 4-trifluoromethoxy benzene-1-thiol, 4-trifluoromethoxythiophenol, benzenethiol, 4-trifluoromethoxy, pubchem15268, acmc-1bxy9, 4-trifluoromethoxy-thiophenol, p-trifluoromethoxybenzenethiol
2777360
4-(trifluoromethoxy)benzenethiol
C1=CC(=CC=C1OC(F)(F)F)S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 64°C to 66°C (15 mmHg)
Refractive Index 1.475
Quantity 1 g
UN Number UN2810
Beilstein 7916918
Sensitivity Air Sensitive
Formula Weight 194.18
Percent Purity 95%
Chemical Name or Material 4-(Trifluoromethoxy)thiophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'