missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenylacetonitrile, 98%, Thermo Scientific™

Catalog Number 11489756
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
49561-96-8
C9H6F3NO
201.15
MFCD00061240
LYFCAROXYJTUQF-UHFFFAOYSA-N
4-trifluoromethoxy phenylacetonitrile, 2-4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzyl cyanide, 4-trifluoromethoxy benzyl cyanide, benzeneacetonitrile, 4-trifluoromethoxy, 4-trifluoromethoxy-phenyl-acetonitrile, 4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzylcyanide, 1-4-trifluoromethoxy phenyl acetonitrile
142676
2-[4-(trifluoromethoxy)phenyl]acetonitrile
FC(F)(F)OC1=CC=C(CC#N)C=C1
This item is not returnable. View return policy
49561-96-8
C9H6F3NO
201.15
MFCD00061240
LYFCAROXYJTUQF-UHFFFAOYSA-N
4-trifluoromethoxy phenylacetonitrile, 2-4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzyl cyanide, 4-trifluoromethoxy benzyl cyanide, benzeneacetonitrile, 4-trifluoromethoxy, 4-trifluoromethoxy-phenyl-acetonitrile, 4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzylcyanide, 1-4-trifluoromethoxy phenyl acetonitrile
142676
2-[4-(trifluoromethoxy)phenyl]acetonitrile
FC(F)(F)OC1=CC=C(CC#N)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 28°C to 30°C
Density 1.28
Flash Point >110°C (230°F)
Refractive Index 1.448
Quantity 1 g
UN Number UN3439
Beilstein 2643789
Formula Weight 201.15
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)phenylacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'