missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)phenylacetonitrile, 98%, Thermo Scientific™

Catalog Number 10659224
Click to view available options
1g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 49561-96-8
CAS Min % 97.5
CAS Max % 100.0
Melting Point 28°C to 30°C
Color Yellow
Density 1.2800g/mL
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H6F3NO
Linear Formula F3COC6H4CH2CN
Refractive Index 1.4480 to 1.45
MDL Number MFCD00061240
Quantity 1 g
Specific Gravity 1.28
Synonym 4-trifluoromethoxy phenylacetonitrile, 2-4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzyl cyanide, 4-trifluoromethoxy benzyl cyanide, benzeneacetonitrile, 4-trifluoromethoxy, 4-trifluoromethoxy-phenyl-acetonitrile, 4-trifluoromethoxy phenyl acetonitrile, 4-trifluoromethoxybenzylcyanide, 1-4-trifluoromethoxy phenyl acetonitrile
InChI Key LYFCAROXYJTUQF-UHFFFAOYSA-N
SMILES FC(F)(F)OC1=CC=C(CC#N)C=C1
IUPAC Name 2-[4-(trifluoromethoxy)phenyl]acetonitrile
Molecular Weight (g/mol) 201.15
PubChem CID 142676
Formula Weight 201.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)phenylacetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethoxy)phenylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'