missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)phenyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 97.0 |
Density | 1.3500g/mL |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | CF3OC6H4NCS |
Quantity | 1 g |
Synonym | 4-trifluoromethoxy phenyl isothiocyanate, 1-isothiocyanato-4-trifluoromethoxy benzene, benzene,1-isothiocyanato-4-trifluoromethoxy, benzene, 1-isothiocyanato-4-trifluoromethoxy, 4-trifluoromethoxy phenylisothiocyanate, 1-isothiocyanato-4-trifluoromethoxy-benzene, acmc-1b5bx, 4-trifluoromethoxyphenyl isothiocyanate, 4-trifluoromethoxyphenyl-isothiocyanate, 4-trifluoromethoxy benzenisothiocyanate |
Formula Weight | 219.19 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Trifluoromethoxy)phenyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'