missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzyl bromide, 98%, Thermo Scientific™

Catalog Number 10702695
Click to view available options
Quantity:
500 mg
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
50824-05-0
C8H6BrF3O
255.03
MFCD00061238
JDNPUJCKXLOHOW-UHFFFAOYSA-N
4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene
142785
1-(bromomethyl)-4-(trifluoromethoxy)benzene
C1=CC(=CC=C1CBr)OC(F)(F)F
This item is not returnable. View return policy
50824-05-0
C8H6BrF3O
255.03
MFCD00061238
JDNPUJCKXLOHOW-UHFFFAOYSA-N
4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene
142785
1-(bromomethyl)-4-(trifluoromethoxy)benzene
C1=CC(=CC=C1CBr)OC(F)(F)F
CAS Min % 97.5
CAS Max % 100.0
Melting Point 23°C to 24°C
Color Yellow
Density 1.5800g/mL
Boiling Point 82°C to 84°C (10.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4CH2Br
Refractive Index 1.4800 to 1.482
Quantity 10 g
Specific Gravity 1.58
Formula Weight 255.03
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-(Trifluoromethoxy)benzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethoxy)benzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'