missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethoxy)benzyl bromide, 98%, Thermo Scientific™
Click to view available options
Quantity:
500 mg
2.5 g
10 g
Packaging:
Glass Bottle
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 23°C to 24°C |
Color | Yellow |
Density | 1.5800g/mL |
Boiling Point | 82°C to 84°C (10.0 mmHg) |
Flash Point | 94°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass Bottle |
Show More |
Safety and Handling
Product Identifier
- 4-(Trifluoromethoxy)benzyl bromide
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'