missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7022608
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
94108-56-2
C7H4ClF3O3S
260.611
MFCD00042408
UHCDBMIOLNKDHG-UHFFFAOYSA-N
4-trifluoromethoxy benzenesulfonyl chloride, 4-trifluoromethoxy benzene-1-sulfonyl chloride, 4-trifluoromethoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethoxy, p-trifluoromethoxy benzenesulphonyl chloride, 4-trifluoromethoxyphenyl-sulfonylchloride, 4-trifluoromethoxyphenylsulfonyl chloride, 4-trifluoromethoxy benzensulfonylchloride, 4-trifluoromethoxy-benzenesulfonylchloride, 4-trifluoromethoxy-benzenesulfonyl chloride
523102
4-(trifluoromethoxy)benzenesulfonyl chloride
C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)Cl
This item is not returnable. View return policy
94108-56-2
C7H4ClF3O3S
260.611
MFCD00042408
UHCDBMIOLNKDHG-UHFFFAOYSA-N
4-trifluoromethoxy benzenesulfonyl chloride, 4-trifluoromethoxy benzene-1-sulfonyl chloride, 4-trifluoromethoxy benzenesulphonyl chloride, benzenesulfonyl chloride, 4-trifluoromethoxy, p-trifluoromethoxy benzenesulphonyl chloride, 4-trifluoromethoxyphenyl-sulfonylchloride, 4-trifluoromethoxyphenylsulfonyl chloride, 4-trifluoromethoxy benzensulfonylchloride, 4-trifluoromethoxy-benzenesulfonylchloride, 4-trifluoromethoxy-benzenesulfonyl chloride
523102
4-(trifluoromethoxy)benzenesulfonyl chloride
C1=CC(=CC=C1OC(F)(F)F)S(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.536
Boiling Point 120°C to 122°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.48
Quantity 1 g
UN Number UN3265
Beilstein 7641524
Sensitivity Moisture sensitive
Formula Weight 260.62
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)benzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'