missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzaldehyde oxime, 97%, Maybridge

Brand:  Maybridge SPB05555DA

Code : 31

Additional Details : CAS Number : 150162-39-3 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10079491

  • £16.00 / 1g


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Specifications

Chemical Identifiers

4-(Trifluoromethoxy)benzaldehyde oxime
97.0
97%
CF3OC6H4CH=NOH
MFCD00210176
150162-39-3
97%
C8H6F3NO2
205.13

Specifications

4-(Trifluoromethoxy)benzaldehyde oxime
150162-39-3
Amber Glass Bottle
97%
CF3OC6H4CH=NOH
MFCD00210176
1g
97.0
97%
C8H6F3NO2
205.13
Videos
SDS