missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzaldehyde, 99%, Thermo Scientific Chemicals

Catalog Number 10549443
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
659-28-9
C8H5F3O2
190.12
MFCD00041530
XQNVDQZWOBPLQZ-UHFFFAOYSA-N
4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
69573
4-(trifluoromethoxy)benzaldehyde
C1=CC(=CC=C1C=O)OC(F)(F)F
This item is not returnable. View return policy
659-28-9
C8H5F3O2
190.12
MFCD00041530
XQNVDQZWOBPLQZ-UHFFFAOYSA-N
4-trifluoromethoxy benzaldehyde, p-trifluoromethoxybenzaldehyde, p-trifluoromethoxy benzaldehyde, benzaldehyde, 4-trifluoromethoxy, 4-trifluoromethoxy-benzaldehyde, alpha,alpha,alpha-trifluoroanisaldehyde, pubchem1471, acmc-209nsx
69573
4-(trifluoromethoxy)benzaldehyde
C1=CC(=CC=C1C=O)OC(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Green-Yellow
Density 1.3300g/mL
Boiling Point 80°C (16.0 mmHg)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3OC6H4CHO
Refractive Index 1.4570 to 1.459
Quantity 50 g
Specific Gravity 1.33
Formula Weight 190.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Trifluoromethoxy)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'